Merck
  • hesperidin induces paraptosis like cell death in hepatoblastoma, HepG2 Cells: involvement of ERK1/2 MAPK [corrected].
hesperidin induces paraptosis like cell death in hepatoblastoma, HepG2 Cells: involvement of ERK1/2 MAPK [corrected].
PloS one (2014-07-01)
Silvia Yumnam, Hyeon Soo Park, Mun Ki Kim, Arulkumar Nagappan, Gyeong Eun Hong, Ho Jeong Lee, Won Sup Lee, Eun Hee Kim, Jae Hyeon Cho, Sung Chul Shin, Gon Sup Kim
ABSTRACT
Hesperidin, a natural flavonoid abundantly present in Citrus is known for its anti-cancer, anti-oxidant and anti-inflammatory properties. In this study we examined the effect of hesperidin on HepG2 cells. HepG2 cells treated with various concentration of hesperidin undergo a distinct type of programed cell death. Cytoplasmic vacuolization, mitochondria and endoplasmic reticulum swelling and uncondensed chromatin were observed in hesperidin treated cells. DNA electrophoresis show lack of DNA fragmentation and western blot analysis demonstrates lack of caspase activation and PARP cleavage. It was observed that hesperidin induced cell death is nonautophagic and also activate mitogen activated protein kinase ERK1/2. Taken together, the data indicate that hesperidin induces paraptosis like cell death in HepG2 cells with the activation of ERK1/2. Thus our finding suggests that hesperidin inducing paraptosis may offer an alternative tool in human liver carcinoma therapy.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Supelco
Hesperidin, analytical standard
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Hesperidin, ≥80% (HPLC)
Hesperidin, European Pharmacopoeia (EP) Reference Standard
Hesperidin, primary reference standard