Merck
  • Snail2/Slug cooperates with Polycomb repressive complex 2 (PRC2) to regulate neural crest development.
Snail2/Slug cooperates with Polycomb repressive complex 2 (PRC2) to regulate neural crest development.
Development (Cambridge, England) (2015-01-27)
Chih-Liang Tien, Amanda Jones, Hengbin Wang, Magda Gerigk, Susan Nozell, Chenbei Chang
ABSTRACT
Neural crest cells arise from the border of the neural plate and epidermal ectoderm, migrate extensively and differentiate into diverse cell types during vertebrate embryogenesis. Although much has been learnt about growth factor signals and gene regulatory networks that regulate neural crest development, limited information is available on how epigenetic mechanisms control this process. In this study, we show that Polycomb repressive complex 2 (PRC2) cooperates with the transcription factor Snail2/Slug to modulate neural crest development in Xenopus. The PRC2 core components Eed, Ezh2 and Suz12 are expressed in the neural crest cells and are required for neural crest marker expression. Knockdown of Ezh2, the catalytic subunit of PRC2 for histone H3K27 methylation, results in defects in neural crest specification, migration and craniofacial cartilage formation. EZH2 interacts directly with Snail2, and Snail2 fails to expand the neural crest domains in the absence of Ezh2. Chromatin immunoprecipitation analysis shows that Snail2 regulates EZH2 occupancy and histone H3K27 trimethylation levels at the promoter region of the Snail2 target E-cadherin. Our results indicate that Snail2 cooperates with EZH2 and PRC2 to control expression of the genes important for neural crest specification and migration during neural crest development.
Product Description
Millipore
Glutathione−Agarose, lyophilized powder
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening