Merck
  • Polyethyleneimine-grafted collagen fiber as a carrier for cell immobilization.
Polyethyleneimine-grafted collagen fiber as a carrier for cell immobilization.
Journal of industrial microbiology & biotechnology (2014-12-30)
Deyi Zhu, Xia Li, Xuepin Liao, Bi Shi
ABSTRACT
Collagen fiber (CF), an abundant natural biopolymer, features many favorable properties that make it a potential carrier for cell immobilization. In the present investigation, CF was grafted with polyethyleneimine (PEI) using glutaraldehyde (GA) as the cross-linking agent, resulting in the formation of a novel CF based carrier (CF-PEI). The properties of CF-PEI as a carrier were evaluated by the immobilization of Microbacterium arborescens (CICC 20196), which has glucose isomerase (EC 5.3.1.5) activity. It was found that M. arborescens cells immobilized on CF-PEI exhibited higher glucose isomerization than those using activated carbon or anion exchange resin as the carriers. The Michaelis constant (K m) of the isomerization reaction for the CF-PEI-immobilized M. arborescens cells was 0.528 mol/L, which was slightly higher than that of free cells (0.473 mol/L). In addition, the apparent activation energies (E a) of free and immobilized cells on CF-PEI were almost the same at 60 kJ/mol. In an isomerization reaction of glucose to fructose in a fixed-bed reactor, CF-PEI-immobilized M. arborescens cells showed appreciable activity and operational stability. The corresponding isomerization ratio was as high as 41 % for 20 days, and the half-life was about 40 days.
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Diallyldimethylammonium chloride, ≥97.0% (AT)
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Diallyldimethylammonium chloride solution, 65 wt. % in H2O