Merck
PAX5 is the transcriptional activator of mucolipin-2 (MCOLN2) gene.
Gene (2014-12-03)
Jessica A Valadez, Math P Cuajungco
ABSTRACT
Transient receptor potential mucolipin (TRPML) proteins belong to the TRP superfamily of non-selective cation channels. The TRPML1, -2, and -3 proteins are encoded by Mucolipin (MCOLN)-1, -2 and -3 genes, respectively. TRPML1 has been associated with mucolipidosis type IV (MLIV), while no disease phenotype has been linked with TRPML2 or -3 protein. The TRPML proteins share high sequence similarities, form hetero-tetramers, and serve in membrane trafficking, autophagy, and metal homeostasis. Previous studies suggest that TRPML2 serves a role in the immune system; however, the evidence is mostly indirect. We hypothesize that if TRPML2 is involved in immune function its expression would be likely regulated by an immune-associated transcription factor protein. Thus, we set out to identify the core promoter region and the transcription factor responsible for MCOLN2 gene expression. Using dual-luciferase assay and over-expression analyses, we reveal for the first time that B-cell lineage specific activator protein (BSAP), also known as paired box 5 (PAX5), controls MCOLN2 expression. Specifically, heterologous expression of PAX5 in HEK-293 cells significantly increased endogenous MCOLN2 transcript and TRPML2 protein levels, while RNA interference targeting endogenous PAX5 reduced its effect. Site-directed mutagenesis studies showed that the core promoter and PAX5 binding region to be between -79 and -60 base pairs upstream of the transcriptional start site. Thus, our findings add to a growing list of evidence for TRPML2's possible involvement in the immune system. The knowledge gained from this study could be used to further characterize the role of TRPML2 in B-cell development and function.
Product Description
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard