Merck
  • Separation of the isomers of benzene poly(carboxylic acid)s by quaternary ammonium salt via formation of deep eutectic solvents.
Separation of the isomers of benzene poly(carboxylic acid)s by quaternary ammonium salt via formation of deep eutectic solvents.
The journal of physical chemistry. B (2014-10-30)
Yucui Hou, Jian Li, Shuhang Ren, Muge Niu, Weize Wu
ABSTRACT
Because of similar properties and very low volatility, isomers of benzene poly(carboxylic acid)s (BPCAs) are very difficult to separate. In this work, we found that isomers of BPCAs could be separated efficiently by quaternary ammonium salts (QASs) via formation of deep eutectic solvents (DESs). Three kinds of QASs were used to separate the isomers of BPCAs, including the isomers of benzene tricarboxylic acids (trimellitic acid, trimesic acid, and hemimellitic acid) and the isomers of benzene dicarboxylic acids (phthalic acid and isophthalic acid). Among the QASs, tetraethylammonium chloride was found to have the best performance, which could completely separate BPCA isomers in methyl ethyl ketone solutions. It was found that the hydrogen bond forming between QAS and BPCA results in the selective separation of BPCA isomers. QAS in DES was regenerated effectively by the antisolvent method, and the regenerated QAS was reused four times with the same high efficiency.
Product Description
Supelco
2-Butanone, analytical standard
Sigma-Aldrich
Isophthalic acid, 99%
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Supelco
Methyl Ethyl Ketone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Supelco
Phthalic acid, analytical standard
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Phthalic acid, European Pharmacopoeia (EP) Reference Standard