Merck
  • Increased prepubertal body weight enhances leptin sensitivity in proopiomelanocortin and neuropeptide y neurons before puberty onset in female rats.
Increased prepubertal body weight enhances leptin sensitivity in proopiomelanocortin and neuropeptide y neurons before puberty onset in female rats.
Endocrinology (2015-01-13)
David Castro-González, Esther Fuente-Martín, Miguel A Sánchez-Garrido, Pilar Argente-Arizón, Manuel Tena-Sempere, Vicente Barrios, Julie A Chowen, Jesús Argente
ABSTRACT
Pubertal onset may be advanced by obesity, with leptin potentially acting as a permissive factor. We hypothesized that having increased body weight (BW) prepubertally affects the ability of leptin to activate intracellular signaling pathways and modulate the expression of hypothalamic neuropeptides involved in reproduction and metabolism. Because being raised in small litters (SLs) tends to increase BW at weaning, female rats were raised in litters of 4 or large litters (LLs) of 12 pups. Leptin (3 μg/g BW) or vehicle (saline) was injected sc at postnatal day (PND) 21 and 30. Rats raised in SLs weighed more at both ages, but relative visceral and subcutaneous fat was increased only on PND21. Serum leptin levels were not different at PND21 or PND30. At PND21, key elements of intracellular leptin signaling (phosphorylated signal transducer and activator of transcription 3 and phosphorylated Akt [p-Akt]) were lower in SL than in LL rats. Leptin injection stimulated phosphorylated signal transducer and activator of transcription 3 in both groups, with a greater increase in LL, whereas p-Akt rose only in SL rats. At PND30, basal leptin signaling did not differ between LL and SL rats. Leptin activation of Akt was similar at 45 minutes, but at 2 hours p-AKT levels were higher in SL than in LL rats, as was the decrease in neuropeptide Y mRNA and increase in pro-opiomelanocortin mRNA levels. No change in the reproductive axis was found. Thus, being raised in SLs increases BW and visceral body fat content, fails to increase plasma leptin concentrations, and increases the leptin responsiveness of both neuropeptide Y and pro-opiomelanocortin cells in the prepubertal hypothalamus.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
SAFC
Sodium deoxycholate