Merck
Investigation of dendriplexes by ion mobility-mass spectrometry.
Molecules (Basel, Switzerland) (2014-12-17)
Emma-Dune Leriche, Marie Hubert-Roux, Carlos Afonso, Catherine M Lange, Martin C Grossel, Florian Maire, Corinne Loutelier-Bourhis
ABSTRACT
Highly branched polyamidoamine (PAMAM) dendrimers presenting biological activities have been envisaged as non-viral gene delivery vectors. They are known to associate with nucleic acid (DNA) in non-covalent complexes via electrostatic interactions. Although their transfection efficiency has been proved, PAMAMs present a significant cytotoxicity due to their cationic surface. To overcome such a drawback, different chemical modifications of the PAMAM surface have been reported such as the attachment of hydrophobic residues. In the present work, we studied the complexation of DNA duplexes with different low-generation PAMAM; ammonia-cored G0(N) and G1(N) PAMAM, native or chemically modified with aromatic residues, i.e., phenyl-modified-PAMAM G0(N) and phenylalanine-modified-PAMAM G1(N). To investigate the interactions involved in the PAMAM/DNA complexes, also called dendriplexes, we used electrospray ionization (ESI) coupled to ion mobility spectrometry-mass-spectrometry (IM-MS). ESI is known to allow the study of non-covalent complexes in native conditions while IM-MS is a bidimensional separation technique particularly useful for the characterization of complex mixtures. IM-MS allows the separation of the expected complexes, possible additional non-specific complexes and the free ligands. Tandem mass spectrometry (MS/MS) was also used for the structural characterization. This work highlights the contribution of IM-MS and MS/MS for the study of small dendriplexes. The stoichiometries of the complexes and the equilibrium dissociation constants were determined. The [DNA/native PAMAM] and [DNA/modified-PAMAM] dendriplexes were compared.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Phenyl isothiocyanate, Sigma Grade, 8.36 M, suitable for solid phase protein sequencing analysis, ≥99% (GC), liquid
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Phenyl isothiocyanate, for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
Sigma-Aldrich
1-Hydroxybenzotriazole hydrate, ≥97.0% dry basis (T)
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
2,2,2-Trifluoroethanol, analytical standard, for NMR spectroscopy
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phenyl isothiocyanate, reagent grade, 98%
Sigma-Aldrich
1-Hydroxybenzotriazole hydrate, wetted with not less than 14 wt. % water, 98% dry basis
Sigma-Aldrich
Phenyl isothiocyanate, 99%, for protein sequencing
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%