Merck
  • Total inhibition of (1)O2-induced oxidative damage to guanine bases of DNA/RNA by turmeric extracts.
Total inhibition of (1)O2-induced oxidative damage to guanine bases of DNA/RNA by turmeric extracts.
Biochemical and biophysical research communications (2014-09-03)
Prakash C Joshi, Hsin H Li, Monique Merchant, Thomas C Keane
ABSTRACT
The guanine base of nucleic acids is known to be very reactive towards degradation by (1)O2-induced oxidative stress. Oxidative reactions of DNA are linked to many human diseases including cancer. Among the various forms of reactive O2 species (OH, (1)O2 or O2(-)), the oxidative stress caused by (1)O2 is of particular physiologic importance because of its selectively long life in aqueous medium and its ability to diffuse through a cell membrane. In this study we investigated the degradation of a model compound guanosine (Guo) by (1)O2, which was generated by riboflavin-induced photosensitization and by molybdate ion catalyzed disproportionation of H2O2. We observed the remarkable ability of an aqueous and alcoholic extracts of Turmeric (Curcuma longa) as an extraordinary scavenger of (1)O2 to completely inhibit the degradation of Guo. The alcoholic extracts were more effective in their antioxidant activity than the corresponding water extract. This naturally occurring antioxidant offers a most economical supplement to protect biologically significant molecules from the oxidative stress induced by (1)O2.
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Octane, anhydrous, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
Dabco® 33-LV
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination