Merck
  • Metal-chloroquine derivatives as possible anti-malarial drugs: evaluation of anti-malarial activity and mode of action.
Metal-chloroquine derivatives as possible anti-malarial drugs: evaluation of anti-malarial activity and mode of action.
Malaria journal (2014-12-05)
Maribel Navarro, William Castro, Marilyn Madamet, Rémy Amalvict, Nicolas Benoit, Bruno Pradines
ABSTRACT
Malaria still has significant impacts on the world; particularly in Africa, South America and Asia where spread over several millions of people and is one of the major causes of death. When chloroquine diphosphate (CQDP) lost its efficiency as a first-line anti-malarial drug, this was a major setback in the effective control of malaria. Currently, malaria is treated with a combination of two or more drugs with different modes of action to provide an adequate cure rate and delay the development of resistance. Clearly, a new effective and non-toxic anti-malarial drug is urgently needed. All metal-chloroquine (CQ) and metal-CQDP complexes were synthesized under N(2) using Schlenk techniques. Their interactions with haematin and the inhibition of β-haematin formation were examined, in both aqueous medium and near water/n-octanol interfaces at pH 5. The anti-malarial activities of these metal- CQ and metal-CQDP complexes were evaluated in vitro against two strains, the CQ-susceptible strain (CQS) 3D7 and the CQ-resistant strain (CQR) W2. The previously synthesized Au(CQ)(Cl) (1), Au(CQ)(TaTg) (2), Pt(CQDP)(2)Cl(2) (3), Pt(CQDP)(2)I(2) (4), Pd(CQ)(2)Cl(2) (5) and the new one Pd(CQDP)(2)I(2) (6) showed better anti-malarial activity than CQ, against the CQS strain; moreover, complexes 2, 3 and 4 were very active against CQR strain. These complexes (1-6) interacted with haem and inhibited β-haematin formation both in aqueous medium and near water/n-octanol interfaces at pH 5 to a greater extent than chloroquine diphosphate (CQDP) and other known metal-based anti-malarial agents. The high anti-malarial activity displayed for these metal-CQ and metal-CQDP complexes (1-6) could be attributable to their effective interaction with haem and the inhibition of β-haematin formation in both aqueous medium and near water/n-octanol interfaces at pH 5.
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
SAFC
HEPES
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%