Merck
  • Neutrophil-associated central nervous system inflammation in tuberculous meningitis immune reconstitution inflammatory syndrome.
Neutrophil-associated central nervous system inflammation in tuberculous meningitis immune reconstitution inflammatory syndrome.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America (2014-08-12)
Suzaan Marais, Katalin A Wilkinson, Maia Lesosky, Anna K Coussens, Armin Deffur, Dominique J Pepper, Charlotte Schutz, Zahiera Ismail, Graeme Meintjes, Robert J Wilkinson
ABSTRACT
The immunopathogenesis of tuberculosis-associated immune reconstitution inflammatory syndrome (IRIS) remains incompletely understood, and we know of only 1 disease site-specific study of the underlying immunology; we recently showed that Mycobacterium tuberculosis culture positivity and increased neutrophils in the cerebrospinal fluid (CSF) of patients with tuberculous meningitis (TBM) are associated with TBM-IRIS. In this study we investigated inflammatory mediators at the disease site in patients with TBM-IRIS. We performed lumbar puncture at 3-5 time points in human immunodeficiency virus (HIV)-infected patients with TBM (n = 34), including at TBM diagnosis, at initiation of antiretroviral therapy (ART) (day 14), 14 days after ART initiation, at presentation of TBM-IRIS, and 14 days thereafter. We determined the concentrations of 40 mediators in CSF (33 paired with blood) with Luminex or enzyme-linked immunosorbent assays. Findings were compared between patients who developed TBM-IRIS (n = 16) and those who did not (TBM-non-IRIS; n = 18). At TBM diagnosis and 2 weeks after ART initiation, TBM-IRIS was associated with severe, compartmentalized inflammation in the CSF, with elevated concentrations of cytokines, chemokines, neutrophil-associated mediators, and matrix metalloproteinases, compared with TBM-non-IRIS. Patients with TBM-non-IRIS whose CSF cultures were positive for M. tuberculosis at TBM diagnosis (n = 6) showed inflammatory responses similar to those seen in patients with TBM-IRIS at both time points. However, at 2 weeks after ART initiation, S100A8/A9 was significantly increased in patients with TBM-IRIS, compared with patients with TBM-non-IRIS whose cultures were positive at baseline. A high baseline M. tuberculosis antigen load drives an inflammatory response that manifests clinically as TBM-IRIS in most, but not all, patients with TBM. Neutrophils and their mediators, especially S100A8/A9, are closely associated with the central nervous system inflammation that characterizes TBM-IRIS.
Sigma-Aldrich
Lamivudine, ≥98% (HPLC), powder
USP
Lamivudine, United States Pharmacopeia (USP) Reference Standard
Supelco
Lamivudine, Pharmaceutical Secondary Standard; Certified Reference Material
Lamivudine, European Pharmacopoeia (EP) Reference Standard
Lamivudine for system suitability 1, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Methyl-2-propanethiol, 99%
Sigma-Aldrich
Efavirenz, ≥98% (HPLC)
Lamivudine for system suitability 2, European Pharmacopoeia (EP) Reference Standard