Merck
  • Mechanism of chemical degradation and determination of solubility by kinetic modeling of the highly unstable sesquiterpene lactone nobilin in different media.
Mechanism of chemical degradation and determination of solubility by kinetic modeling of the highly unstable sesquiterpene lactone nobilin in different media.
Journal of pharmaceutical sciences (2014-09-02)
Ursula Thormann, Maria De Mieri, Markus Neuburger, Sheela Verjee, Peter Altmann, Matthias Hamburger, Georgios Imanidis
ABSTRACT
The objective of this work was first to investigate the chemical degradation of the sesquiterpene lactone nobilin and determine its solubility under conditions of concurrent degradation for partially amorphous starting material; second, to determine the effect of biorelevant media used in the in vitro measurement of intestinal absorption on degradation and solubility of nobilin. Purely aqueous medium (aq-TMCaco ), fasted and fed state simulated intestinal fluid (FaSSIF-TMCaco and FeSSIF-TMCaco ), and two liposomal formulations (LiposomesFaSSIF and LiposomesFeSSIF ) with the same lipid concentration as FaSSIF-TMCaco and FeSSIF-TMCaco were used. Degradation products were identified by nuclear magnetic resonance and X-ray crystallography and the order of reaction kinetics was determined. Solubility was deduced with a mathematical model encompassing dissolution, degradation, and reprecipitation kinetics that took into account particle size distribution of the solid material. Degradation mechanism of nobilin involved water-catalyzed opening of the lactone ring and transannular cyclization resulting in five degradation products. Degradation followed first-order kinetics in aq-TMCaco and FaSSIF-TMCaco , and higher-order kinetics in FeSSIF-TMCaco and the two liposomal formulations, whereas degradation in the latter media was diminished. Solubility of nobilin increased in the order: aq-TMCaco < FaSSIF-TMCaco , < LiposomesFaSSIF < FeSSIF-TMCaco < LiposomesFeSSIF . Improvement of stability and solubility was consistent with the incorporation of the nobilin molecule into colloidal lipid particles. The developed kinetic model is proposed to be a useful tool for deducing solubility under dynamic conditions.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Ethylene sulfite, 98%
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)