Merck
  • Physical and dissolution characterization of cilostazol solid dispersions prepared by hot melt granulation (HMG) and thermal adhesion granulation (TAG) methods.
Physical and dissolution characterization of cilostazol solid dispersions prepared by hot melt granulation (HMG) and thermal adhesion granulation (TAG) methods.
International journal of pharmaceutics (2014-08-05)
Ying-Chen Chen, Hsiu-O Ho, Jiun-Da Chiou, Ming-Thau Sheu
ABSTRACT
A growing number of poorly water-soluble drug have been discovered, but the poor bioavailability is a critical problem. In this study, physical properties and dissolution profiles of cilostazol solid dispersions prepared by hydrophilic/lipophilic excipients (Kollidon(®) VA64, tocopheryl polyethylene glycol succinate (TPGS), vitamine E) with hot-melt and thermal adhesion granulation (TAG) method to adsorb Fujicalin(®) and Microcel(®) were characterized. Results demonstrate the angle of repose in formulations with Fujicalin(®) was improved than those with Microcel(®), but the difference disappeared when more TPGS or vitamin E was added. Compared the formulation made by hot-melt and TAG method, both improved flowability. The hardness decreased with the increased amount of TPGS and vitamin E. The formulations with Microcel(®) had lower hardness than those with Fujicalin(®), because Microcel(®) has weaker adsorption ability and cannot afford much TPGS and vitamin E, leading to lower hardness. Furthermore, the solubility was almost three-fold higher than that of Pletaal(®) (7.68 ± 0.20 μg/mL) in compositions containing TPGS and vitamin E made by hot-melt or TAG method, in which a controlled drug release pattern was demonstrated. There is no significant difference on dissolution profile between hot-melt and TAG method. However, the procedure of TAG is easier, indicating its potential pharmaceutical use.
Sigma-Aldrich
Cilostazol, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
(±)-α-Tocopherol, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Cilostazol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
USP
Alpha Tocopherol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
α-Tocopherol, European Pharmacopoeia (EP) Reference Standard