Merck
  • Spatial control of Cdc42 signalling by a GM130-RasGRF complex regulates polarity and tumorigenesis.
Spatial control of Cdc42 signalling by a GM130-RasGRF complex regulates polarity and tumorigenesis.
Nature communications (2014-09-12)
Francesco Baschieri, Stefano Confalonieri, Giovanni Bertalot, Pier Paolo Di Fiore, Wolfgang Dietmaier, Marcel Leist, Piero Crespo, Ian G Macara, Hesso Farhan
ABSTRACT
The small GTPase Cdc42 is a key regulator of polarity, but little is known in mammals about its spatial regulation and the relevance of spatial Cdc42 pools for polarity. Here we report the identification of a GM130-RasGRF complex as a regulator of Cdc42 at the Golgi. Silencing GM130 results in RasGRF-dependent inhibition of the Golgi pool of Cdc42, but does not affect Cdc42 at the cell surface. Furthermore, active Cdc42 at the Golgi is important to sustain asymmetric front-rear Cdc42-GTP distribution in directionally migrating cells. Concurrent to Cdc42 inhibition, silencing GM130 also results in RasGRF-dependent Ras-ERK pathway activation. Moreover, depletion of GM130 is sufficient to induce E-cadherin downregulation, indicative of a loss in cell polarity and epithelial identity. Accordingly, GM130 expression is frequently lost in colorectal and breast cancer patients. These findings establish a previously unrecognized role for a GM130-RasGRF-Cdc42 connection in regulating polarity and tumorigenesis.
Sigma-Aldrich
MISSION® esiRNA, targeting human HRAS
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
SAFC
HEPES
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Hras1
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
MISSION® esiRNA, targeting human DNMBP
Sigma-Aldrich
MISSION® esiRNA, targeting human KRAS
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Dnmbp