Merck
  • Functional interplay between Parkin and Drp1 in mitochondrial fission and clearance.
Functional interplay between Parkin and Drp1 in mitochondrial fission and clearance.
Biochimica et biophysica acta (2014-06-01)
Lori Buhlman, Maria Damiano, Giulia Bertolin, Rosa Ferrando-Miguel, Anne Lombès, Alexis Brice, Olga Corti
ABSTRACT
Autosomal recessive early-onset Parkinson's disease is most often caused by mutations in the genes encoding the cytosolic E3 ubiquitin ligase Parkin and the mitochondrial serine/threonine kinase PINK1. Studies in Drosophila models and mammalian cells have demonstrated that these proteins regulate various aspects of mitochondrial physiology, including organelle transport, dynamics and turnover. How PINK1 and Parkin orchestrate these processes, and whether they always do so within a common pathway remain to be clarified. We have revisited the role of PINK1 and Parkin in mitochondrial dynamics, and explored its relation to the mitochondrial clearance program controlled by these proteins. We show that PINK1 and Parkin promote Drp1-dependent mitochondrial fission by mechanisms that are at least in part independent. Parkin-mediated mitochondrial fragmentation was abolished by treatments interfering with the calcium/calmodulin/calcineurin signaling pathway, suggesting that it requires dephosphorylation of serine 637 of Drp1. Parkinson's disease-causing mutations with differential impact on mitochondrial morphology and organelle degradation demonstrated that the pro-fission effect of Parkin is not required for efficient mitochondrial clearance. In contrast, the use of Förster energy transfer imaging microscopy revealed that Drp1 and Parkin are co-recruited to mitochondria in proximity of PINK1 following mitochondrial depolarization, indicating spatial coordination between these events in mitochondrial degradation. Our results also hint at a major role of the outer mitochondrial adaptor MiD51 in Drp1 recruitment and Parkin-dependent mitophagy. Altogether, our observations provide new insight into the mechanisms underlying the regulation of mitochondrial dynamics by Parkin and its relation to the mitochondrial clearance program mediated by the PINK1/Parkin pathway.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Ethopropazine hydrochloride, ≥98% (HPLC), powder
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SAFC
L-Glutamine
Supelco
Forskolin, analytical standard
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland