Merck
  • Stability indicating MEKC method for the determination of gliclazide and its specified impurities.
Stability indicating MEKC method for the determination of gliclazide and its specified impurities.
Journal of pharmaceutical and biomedical analysis (2014-09-27)
Athiporn Doomkaew, Brompoj Prutthiwanasan, Leena Suntornsuk
ABSTRACT
A stability indicating-micellar electrokinetic chromatography method was developed and validated for the determination of gliclazide (GCZ) and its specified impurities, gliclazide impurities B (GZB) and F (GZF) in bulk and tablets. The analytes were well separated (Rs>2.1) in 5 min using 10mM phosphate buffer (pH 7.0) containing 15 mM sodium dodecyl sulfate on a fused-silica capillary with an effective length of 40 cm and an inner diameter of 50 μm, injection at 50 mbar for 5s, temperature of 25°C, applied voltage of 20 kV and photodiode array detection at 225 nm. The method showed good linearity (r(2)>0.99, in the ranges of 128-192, 20-60 and 10-50 μg/mL for GCZ, GZB and GZF, respectively) and precision (%RSD for intra- and inter-day precision of less than 2.00%, n=3) for all compounds. Accuracy represented as %recovery was between 99.1 and 100.1% with %RSDs of less than 0.59% (n=3). Limits of detection and quantitation were less than 40 and 120 μg/mL, respectively. The method was robust with %RSDs of migration time and peak areas of less than 1.36% (n=9), when buffer and separating voltage were altered around the optimal values. Stress tests showed that GCZ was stable in alkaline hydrolysis both at room and elevated temperature. However, GCZ degraded under acid and neutral hydrolysis and oxidation condition. Elevated temperature and exposure to sunlight accelerated GCZ degradation and formation of GZB and an unknown degradation product. Stability profiles and degradation kinetics of GCZ could be established using the MEKC method. In addition, the method could be used for assay of GCZ in raw material and commercial tablets and results revealed that contents of GCZ in all samples were within the pharmacopeia limit. No degradation products, especially GZB and GZF, were observed in the investigated samples.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%