Merck
  • Biodegradation of fenoxaprop-P-ethyl (FE) by Acinetobacter sp. strain DL-2 and cloning of FE hydrolase gene afeH.
Biodegradation of fenoxaprop-P-ethyl (FE) by Acinetobacter sp. strain DL-2 and cloning of FE hydrolase gene afeH.
Bioresource technology (2015-03-31)
Weiliang Dong, Sheng Jiang, Kaiwen Shi, Fei Wang, Shuhuan Li, Jie Zhou, Fei Huang, Yicheng Wang, Yuxiao Zheng, Ying Hou, Yan Huang, Zhongli Cui
ABSTRACT
Fenoxaprop-P-ethyl (FE) is widely used as a post-emergence aryloxyphenoxy propionate (AOPP) herbicide in agriculture. An efficient FE-degrading strain DL-2 was isolated from the enrichment culture and identified as Acinetobacter sp. and the metabolite fenoxaprop acid (FA) was identified by HPLC/MS analysis. The strain DL-2 could also degrade a wide range of other AOPP herbicides. A novel FE hydrolase esterase gene afeH was cloned from strain DL-2 and functionally expressed in Escherichia coli BL21(DE3). The specific activities of recombinant AfeH was 216.39 U mg(-1) for FE with Km and Vmax values of 0.82 μM and 7.94 μmol min(-1) mg(-1). AfeH could also hydrolyze various AOPP herbicides, p-nitrophenyl esters and triglycerides. The optimal pH and temperature for recombinant AfeH were 9.0 and 50°C, respectively; the enzyme was activated by Co(2+) and inhibited by Ca(2+), Zn(2+), Ba(2+). AfeH was inhibited strongly by phenylmethylsulfonyl and SDS and weakly by dimethyl sulfoxide.
Sigma-Aldrich
Hydriodic acid, distilled, 57 wt. %, 99.999% trace metals basis
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, ACS reagent, 55%
Sigma-Aldrich
Hydriodic acid, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%
Imidazole, European Pharmacopoeia (EP) Reference Standard
Supelco
6-Benzylaminopurine, PESTANAL®, analytical standard
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
6-Benzylaminopurine, suitable for plant cell culture
Sigma-Aldrich
Nitrilotriacetic acid trisodium salt, Sigma Grade, ≥98%
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
6-Benzylaminopurine, ReagentPlus®, ≥99.0% (HPLC)
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri