Merck
  • Knockdown of c-Myc inhibits cell proliferation by negatively regulating the Cdk/Rb/E2F pathway in nasopharyngeal carcinoma cells.
Knockdown of c-Myc inhibits cell proliferation by negatively regulating the Cdk/Rb/E2F pathway in nasopharyngeal carcinoma cells.
Acta biochimica et biophysica Sinica (2015-01-30)
Zhaoxia Niu, Huaying Liu, Ming Zhou, Heran Wang, Yukun Liu, Xiayu Li, Wei Xiong, Jian Ma, Xiaoling Li, Guiyuan Li
ABSTRACT
The proto-oncogene c-Myc encodes a transcription factor that is involved in the regulation of cellular proliferation, differentiation, and apoptosis. Several studies indicate that the over-expression of c-Myc is a frequent genetic abnormality in nasopharyngeal carcinoma (NPC). Therefore, specifically reducing its level by genetic means in established NPC cell lines helps to better understand its role in the pathogenesis of NPC. In this study, for the first time, we successfully established and characterized NPC 5-8F cell line with stably suppressed c-Myc expression by employing a DNA-based RNA interference approach. The suppression of c-Myc resulted in reduced cell growth, colony formation, and cell cycle progression in 5-8F cells. In vivo tumor formation assays revealed that the knockdown of c-Myc reduced the tumorigenic potential of 5-8F cells in nude mice. At the molecular level, we found that the knockdown of c-Myc could decrease the expression of several critical molecules involved in the Cdk/Rb/E2F pathway, including CDK4, cyclin D1, CDK2, pRb, E2F3, and DP2, and significantly reduced the promoter activity of cyclin D1. Taken together, these findings provide valuable mechanistic insights into the role of c-Myc in nasopharyngeal carcinogenesis and suggest that the knockdown of c-Myc may be a potential therapeutic approach for the treatment of NPC.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Anti-c-myc Antibody, clone 3C7, clone 3C7, Chemicon®, from mouse
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid