Merck
  • Immunomodulation of RAW 264.7 murine macrophage functions and antioxidant activities of 11 plant extracts.
Immunomodulation of RAW 264.7 murine macrophage functions and antioxidant activities of 11 plant extracts.
Immunological investigations (2015-01-08)
Mohammed Ghonime, Mohamed Emara, Riham Shawky, Hesham Soliman, Ramadan El-Domany, Ahmed Abdelaziz
ABSTRACT
A group of 11 medicinal plants, including Lavandula pubescens, Trigonella foenugricium, Salsola schweinforthi, Calligonum comosum, Silene succulenta, Silene villosa, Bogonvillea glabra, Cakile maritime, Gomphrene celesoids, Mirabilis jalaba, and Silene nocturna growing in Egypt, were extracted and examined for their immunomodulatory and antioxidant activities. RAW 264.7 cells were recruited to investigate the immunomodulatory effect through multiple parameters analysis. First, the proliferation index of macrophages cells was evaluated revealing that Trigonella foenugricium, Silene succulenta and Silene villosa have a significant cytotoxic effect on RAW cells. Interestingly, we observed enhancement of macrophages phagocytic function of by all extracts except Cakile maritime, Gomphrena celosioides and Silene nocturna. Afterwards, macrophages were challenged by incubation with LPS and the effect of various extracts on inflammatory responses was investigated; the generation of NO from activated macrophage was substantially suppressed by 7 extracts namely, Trigonella foenugricium, Calligonum comosum, Silene succulenta, Bougainvillea glabra, Mirabilis jalaba, Gomphrena celosioides and Silene nocturna. TNF-α was decreased by percentage range from 3.8 to 85.8% and Trigonella foenugricium extract showed the highest inhibition of TNF-α release. All extracts except Trigonella foenugricium, Salsola schweinforthi, Silene succulenta and Mirabilis jalaba significantly inhibited COX-2 production from stimulated macrophage. Moreover, evaluating the potential antioxidant activity of these extracts showed that Trigonella foenugricium, Salsola schweinforthi, Calligonum comosum, Bogonvillea glabra and Mirabilis jalaba exhibited some antioxidant activities. Taken together, our results suggest that some of these extracts may have a considerable antinflammatory and antioxidant effects and may be a potential therapeutic choice in the treatment of inflammatory diseases.
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland