Merck
  • The liver-selective NO donor, V-PYRRO/NO, protects against liver steatosis and improves postprandial glucose tolerance in mice fed high fat diet.
The liver-selective NO donor, V-PYRRO/NO, protects against liver steatosis and improves postprandial glucose tolerance in mice fed high fat diet.
Biochemical pharmacology (2014-12-24)
Edyta Maslak, Piotr Zabielski, Kamila Kochan, Kamil Kus, Agnieszka Jasztal, Barbara Sitek, Bartosz Proniewski, Tomasz Wojcik, Katarzyna Gula, Agnieszka Kij, Maria Walczak, Małgorzata Baranska, Adrian Chabowski, Ryan J Holland, Joseph E Saavedra, Larry K Keefer, Stefan Chlopicki
ABSTRACT
There is an unmet medical need for novel NAFLD treatments. Here we have examined the effects of liver-selective NO donor (V-PYRRO/NO) as compared with metformin on hepatic steatosis and glucose tolerance in mice fed high fat diet. Effects of V-PYRRO/NO (5 mgkg(-1)) or metformin (616 mgkg(-1)) were examined in C57BL/6J mice fed high fat diet (HF, 60 kcal% fat). Quantitative determination of steatosis, liver fatty acid composition and western blot analysis of selected proteins involved in mitochondrial biogenesis, fatty acid de novo synthesis and oxidation, triacylglycerols and cholesterol transport from the liver were performed. Liver NOx and nitrate concentration and blood biochemistry were also analyzed. V-PYRRO/NO and metformin reduced liver steatosis with simultaneous reduction of total liver triacylglycerols, diacylglycerols and ceramides fraction and reversed HF-induced decrease in UFA/SFA ratio. V-PYRRO/NO substantially improved postprandial glucose tolerance, while the effect of metformin was modest and more pronounced on HOMA IR index. The anti-steatotic mechanism of V-PYRRO/NO was dependent on NO release, differed from that of metformin and involved improved glucose tolerance and inhibition of de novo fatty acid synthesis by Akt activation and ACC phosphorylation. In turn, major mechanism of metformin action involved increased expression of proteins implicated in mitochondrial biogenesis and metabolism (PGC-1α, PPARα, COX IV, cytochrome c, HADHSC). V-PYRRO/NO acts as a liver-specific NO donor prodrug affording pronounced anti-steatotic effects and may represent an efficient, mechanistically novel approach to prevent liver steatosis and insulin resistance.
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Glucose solution, BioUltra, for molecular biology, ~20% in H2O
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%