Merck
  • Desulfovibrio desulfuricans isolates from the gut of a single individual: structural and biological lipid A characterization.
Desulfovibrio desulfuricans isolates from the gut of a single individual: structural and biological lipid A characterization.
FEBS letters (2014-12-06)
Wei Zhang-Sun, Luis A Augusto, Liping Zhao, Martine Caroff
ABSTRACT
The levels of sulfate-reducing bacteria (SRB), including Desulfovibrionaceae, in the gut increase following a fat-enriched diet. Endotoxins from gut microbiota contribute to the inflammation process, leading to metabolic diseases. Thus, we sought to characterize the lipid A structures of Desulfovibrionaceae lipopolysaccharides (LPS) that are associated with the microbiota inflammatory properties. LPS variants were obtained from two SRB isolates from the gut of a single individual. These LPS variants shared similar lipid A moieties with Enterobacterial LPS, but differed from one another with regard to fatty-acid numbers and endotoxic activity. This first complete structural characterization of Desulfovibrio lipid A gives new insights into previously published data on Desulfovibrio lipid A biosynthesis. LPS microdiversity within SRBs illustrates how adaptation can influence pro-inflammatory potential.
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
2,3-Dihydroxybenzoic acid, 99%
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Water, for TOC analysis
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, Absolute - Acetone free