Merck
  • Supermolecular evodiamine loaded water-in-oil nanoemulsions: enhanced physicochemical and biological characteristics.
Supermolecular evodiamine loaded water-in-oil nanoemulsions: enhanced physicochemical and biological characteristics.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-06-26)
Jiangbo Hu, Lili Sun, Dezhang Zhao, Liangke Zhang, Mengliang Ye, Qunyou Tan, Chunshu Fang, Hong Wang, Jingqing Zhang
ABSTRACT
The purpose of this study was to develop and evaluate the supermolecular evodiamine (EVO) loaded water-in-oil nanoemulsions containing brucea javanica oil (NESEB) with enhanced physicochemical and biological characteristics. NESEB was fabricated by applying supermolecular phytosome nanotechnology and nanoemulsification technology together, in addition to using synergistic plant essential oil as a basic composition. Preferred physicochemical and biological characteristics of NESEB were investigated and compared with free EVO and other nanoemulsive EVO carriers. The possible explanations for improved absorption and bioavailability were put forward here. NESEB had high absorption and bioavailability, for example: the absorption rate constants and permeabilities of NESEB in different intestinal segments were 3.65-6.76 times that of free EVO; the relative bioavailability of NESEB to free EVO was 846.97%. NESEB markedly improved the oral bioavailability of EVO, which was most likely due to the increased gastrointestinal absorption. The development of nanoemulsion-based supermolecular EVO nanocarriers provides valuable tactics in insoluble natural antitumor drug delivering.
Sigma-Aldrich
Ethyl oleate, technical grade, 70%
Sigma-Aldrich
Ethyl oleate, natural, ≥85%
Sigma-Aldrich
Ethyl oleate, 98%
Sigma-Aldrich
Ethyl oleate, tested according to Ph. Eur.
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Honokiol, ≥98% (HPLC), powder
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Supelco
Honokiol, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Honokiol, European Pharmacopoeia (EP) Reference Standard