Merck
  • Nutrients value and antioxidant content of indigenous vegetables from Southern Thailand.
Nutrients value and antioxidant content of indigenous vegetables from Southern Thailand.
Food chemistry (2014-12-04)
Ratchanee Kongkachuichai, Rin Charoensiri, Kameelah Yakoh, Aurawan Kringkasemsee, Poonsub Insung
ABSTRACT
Evidence from epidemiological studies has strongly suggested that diets rich in fruits and vegetables play a vital role in disease prevention. The aim of this study was to determine nutrient and antioxidant content for 15 varieties of indigenous vegetables and fruits collected from Southern Thailand. The data indicated that indigenous vegetables provided small to moderate amounts of macronutrients and minerals. The highest content of β-carotene was found in Indian lettuce (Lactuca indica; 3575.54 μg/100 g), whereas water dropwort (Oenanthe javanica; 7439.11 μg/100 g) had the highest lutein content. Ripe cashew apple (Anacardium occidentale; 178.34 mg/100 g) and Spanish joint fir (Gnetum gnemon; 109.43 mg/100 g) were excellent sources of vitamin C. Mon-pu (Glochidion perakense) and young cashew leaves (Anacardium occidentale) were rich sources of β-carotene, lutein, total polyphenol, especially gallic acid, and had relatively high ORAC and FRAP activities. In conclusion, Thai indigenous vegetables provide diverse natural bioactive compounds that may contribute health benefits to the consumer.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Millipore
2,3,5-Triphenyl-tetrazolium chloride solution, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
(±)-α-Tocopherol, analytical standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
tert-Butylhydroquinone, 97%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline