Merck
  • Comparing different sterol containing solid lipid nanoparticles for targeted delivery of quercetin in hepatocellular carcinoma.
Comparing different sterol containing solid lipid nanoparticles for targeted delivery of quercetin in hepatocellular carcinoma.
Journal of liposome research (2013-12-21)
Jaleh Varshosaz, Abbas Jafarian, Golnaz Salehi, Behzad Zolfaghari
ABSTRACT
Quercetin (QT) is a potential chemotherapeutic drug with low solubility that seriously limits its clinical use. The aim of this study was enhancing cellular penetration of QT by sterol containing solid lipid nanoparticles (SLNs) which make bilayers fluent for targeting hepatocellular carcinoma cells. Three variables including sterol type (cholesterol, stigmasterol and stigmastanol), drug and sterol content were studied in a surface response D-optimal design for preparation of QT-SLNs by emulsification solvent evaporation method. The studied responses included particle size, zeta potential, drug loading capacity and 24 h release efficiency (RE24%). Scanning electron and atomic force microscopy were used to study the morphology of QT-SLNs and their thermal behavior was studied by DSC analysis. Cytotoxicity of QT-SLNs was determined by MTT assay on HepG-2 cells and cellular uptake by fluorescence microscopy method. Optimized QT-SLNs obtained from cholesterol and QT with the ratio of 2:1 that showed particle size of 78.0 ± 7.0 nm, zeta potential of -22.7 ± 1.3 mV, drug loading efficiency of 99.9 ± 0.5% and RE24 of 56.3 ± 3.4%. IC50 of QT in cholesterol SLNs was about six and two times less than free QT and phytosterol SLNs, respectively, and caused more accumulation of QT in HepG2 cells. Blank phytosterol SLNs were toxic on cells.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Chloroform, analytical standard
Supelco
Acetone, analytical standard
Pyrene, BCR®, certified reference material
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
1-Hexadecanol, ≥99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
1-Hexadecanol, 95%