Merck
  • The antiproliferative response of indole-3-carbinol in human melanoma cells is triggered by an interaction with NEDD4-1 and disruption of wild-type PTEN degradation.
The antiproliferative response of indole-3-carbinol in human melanoma cells is triggered by an interaction with NEDD4-1 and disruption of wild-type PTEN degradation.
Molecular cancer research : MCR (2014-07-11)
Ida Aronchik, Aishwarya Kundu, Jeanne G Quirit, Gary L Firestone
ABSTRACT
Human melanoma cells displaying distinct PTEN genotypes were used to assess the cellular role of this important tumor-suppressor protein in the antiproliferative response induced by the chemopreventative agent indole-3-carbinol (I3C), a natural indolecarbinol compound derived from the breakdown of glucobrassicin produced in cruciferous vegetables such as broccoli and Brussels sprouts. I3C induced a G1-phase cell-cycle arrest and apoptosis by stabilization of PTEN in human melanoma cells that express wild-type PTEN, but not in cells with mutant or null PTEN genotypes. Importantly, normal human epidermal melanocytes were unaffected by I3C treatment. In wild-type PTEN-expressing melanoma xenografts, formed in athymic mice, I3C inhibited the in vivo tumor growth rate and increased PTEN protein levels in the residual tumors. Mechanistically, I3C disrupted the ubiquitination of PTEN by NEDD4-1 (NEDD4), which prevented the proteasome-mediated degradation of PTEN without altering its transcript levels. RNAi-mediated knockdown of PTEN prevented the I3C-induced apoptotic response, whereas knockdown of NEDD4-1 mimicked the I3C apoptotic response, stabilized PTEN protein levels, and downregulated phosphorylated AKT-1 levels. Co-knockdown of PTEN and NEDD4-1 revealed that I3C-regulated apoptotic signaling through NEDD4-1 requires the presence of the wild-type PTEN protein. Finally, in silico structural modeling, in combination with isothermal titration calorimetry analysis, demonstrated that I3C directly interacts with purified NEDD4-1 protein. This study identifies NEDD4-1 as a new I3C target protein, and that the I3C disruption of NEDD4-1 ubiquitination activity triggers the stabilization of the wild-type PTEN tumor suppressor to induce an antiproliferative response in melanoma. Mol Cancer Res; 12(11); 1621-34. ©2014 AACR.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture