Merck
  • Restoration of the insulinotropic effect of glucose-dependent insulinotropic polypeptide contributes to the antidiabetic effect of dipeptidyl peptidase-4 inhibitors.
Restoration of the insulinotropic effect of glucose-dependent insulinotropic polypeptide contributes to the antidiabetic effect of dipeptidyl peptidase-4 inhibitors.
Diabetes, obesity & metabolism (2014-09-23)
K Aaboe, S Akram, C F Deacon, J J Holst, S Madsbad, T Krarup
ABSTRACT
To examine whether 12 weeks of treatment with a dipeptidyl peptidase-4 (DPP-4) inhibitor, sitagliptin, influences the insulin secretion induced by glucose, glucose-dependent insulinotropic polypeptide (GIP) and glucagon-like peptide-1 (GLP-1) during a hyperglycaemic clamp in patients with type 2 diabetes (T2DM). A randomized, double-blind, placebo-controlled study was conducted over 12 weeks, during which 25 patients with T2DM completed treatment with either sitagliptin (100 mg once daily) or placebo as add-on therapy to metformin [sitagliptin group (n = 12): mean ± standard error of the mean (s.e.m.) age 54 ± 2.5 years, mean ± s.e.m. HbA1c 7.8 ± 0.2%; placebo group (n = 13): mean ± s.e.m. age: 57 ± 3.0 years, mean ± s.e.m. HbA1c 7.9 ± 0.2 %]. In weeks 1 and 12, the patients underwent three 2-h 15-mM hyperglycaemic clamp experiments with infusion of either saline, GLP-1 or GIP. β-cell function was evaluated according to first-phase, second-phase, incremental and total insulin and C-peptide responses. In the sitagliptin group, the mean HbA1c concentration was significantly reduced by 0.9% (p = 0.01). The total β-cell response during GIP infusion improved significantly from week 1 to week 12, both within the sitagliptin group (p = 0.004) and when compared with the placebo group (p = 0.04). The total β-cell response during GLP-1 infusion was significantly higher (p = 0.001) when compared with saline and GIP infusion, but with no improvement from week 1 to week 12. No significant changes in β-cell function occurred in the placebo group. Treatment with the DPP-4 inhibitor sitagliptin over 12 weeks in patients with T2DM partially restored the lost insulinotropic effect of GIP, whereas the preserved insulinotropic effect of GLP-1 was not further improved. A gradual enhancement of the insulinotropic effect of GIP, therefore, possibly contributes to the antidiabetic actions of DPP-4 inhibitors.
Product Description
Valine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
DL-Valine, ≥97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
DL-Valine, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)