Merck
  • The role of methanol addition to water samples in reducing analyte adsorption and matrix effects in liquid chromatography-tandem mass spectrometry.
The role of methanol addition to water samples in reducing analyte adsorption and matrix effects in liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2015-03-10)
Wei Li, Yucan Liu, Jinming Duan, Christopher P Saint, Dennis Mulcahy
ABSTRACT
Liquid chromatography-tandem mass spectrometry (LC-MS/MS) analysis coupled simply with water filtering before injection has proven to be a simple, economic and time-saving method for analyzing trace-level organic pollutants in aqueous environments. However, the linearity, precision and detection limits of such methods for late-eluting analytes were found to be much poorer than for early-eluting ones due to adsorption of the analytes in the operating system, such as sample vial, flow path and sample loop, creating problems in quantitative analysis. Addition of methanol (MeOH) into water samples as a modifier was shown to be effective in alleviating or even eliminating the negative effect on signal intensity for the late-eluting analytes and at the same time being able to reduce certain matrix effects for real water samples. Based on the maximum detection signal intensity obtained on desorption of the analytes with MeOH addition, the ratio of the detection signal intensity without addition of MeOH to the maximum intensity can be used to evaluate the effectiveness of methanol addition. Accordingly, the values of <50%, 50-80%, 80-120% could be used to indicate strong, medium and no effects, respectively. Based on this concept, an external matrix-matched calibration method with the addition of MeOH has been successfully established for analyzing fifteen pesticides with diverse physico-chemical properties in surface and groundwater with good linearity (r(2): 0.9929-0.9996), precision (intra-day relative standard deviation (RSD): 1.4-10.7%, inter-day RSD: 1.5-9.4%), accuracy (76.9-126.7%) and low limits of detection (0.003-0.028μg/L).
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Malathion, certified reference material, TraceCERT®
Supelco
Malathion, PESTANAL®, analytical standard
Supelco
Diuron, PESTANAL®, analytical standard
Supelco
Propazine, PESTANAL®, analytical standard
Supelco
Fenobucarb, PESTANAL®, analytical standard
Sigma-Aldrich
Diuron, ≥98%
Supelco
Isoproturon, PESTANAL®, analytical standard
Supelco
Profenofos, PESTANAL®, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
USP
Malathion, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Malathion, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Diazinon, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Atrazine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Simazine, PESTANAL®, analytical standard
Supelco
Carbaryl, PESTANAL®, analytical standard
Supelco
Atrazine, analytical standard