Merck
  • Determination of polyphenolic compounds in Cirsium palustre (L.) extracts by high performance liquid chromatography with chemiluminescence detection.
Determination of polyphenolic compounds in Cirsium palustre (L.) extracts by high performance liquid chromatography with chemiluminescence detection.
Talanta (2014-12-02)
Edyta Nalewajko-Sieliwoniuk, Julita Malejko, Monika Mozolewska, Elżbieta Wołyniec, Jolanta Nazaruk
ABSTRACT
The first method for the simultaneous determination of polyphenolic antioxidants in extracts from leaves of Cirsium palustre based on high performance liquid chromatography combined with flow injection chemiluminescence detection (HPLC-FI-CL) has been developed. The extracts were prepared by using methanol as extraction medium and two types of extraction methods (reflux and ultrasound assisted extraction). The post-column CL determination of polyphenols was based on their enhancing effect on the chemiluminescence intensity generated in manganese(IV)-hexametaphosphate-formaldehyde system in a phosphoric acid medium. Main antioxidants determined in C. palustre leaves were eriodictyol-7-O-glucoside, luteolin-7-O-glucoside and 6-hydroxyluteolin-7-O-glucoside belonging to flavonoids, and chlorogenic acid belonging to phenolic acids. Chromatographic separation was carried out on a C18 column with gradient elution by using a mobile phase containing 0.25% (v/v) phosphoric acid in water (solvent A) and 100% methanol (solvent B). Under the optimized conditions of chromatographic separation and CL detection the validation of the method was performed. The calibration curves showed good linearity in the concentration range from 0.5 to 40 µg mL(-1). The HPLC-FI-CL method was successfully applied to the determination of four polyphenolic compounds in methanolic extracts from leaves of C. palustre. The accuracy of the developed method was confirmed by the comparison of the results with those obtained by an HPLC-PDA method. The relative error of determination does not exceed 6.1%. However, the HPLC-FI-CL method is characterized by 40-65 times higher sensitivity compared to the HPLC-PDA method.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, NMR reference standard
Chlorogenic acid, primary reference standard
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Manganese(IV) oxide, activated, ~85%, <10 μm
Sigma-Aldrich
Manganese(IV) oxide, 10 μm, reagent grade, ≥90%