Merck
  • Quercetin and omega 3 ameliorate oxidative stress induced by aluminium chloride in the brain.
Quercetin and omega 3 ameliorate oxidative stress induced by aluminium chloride in the brain.
Journal of molecular neuroscience : MN (2014-02-04)
Haytham Abdallah Ali, Mohamed Afifi, Aaser Mohamed Abdelazim, Yahia Youssef Mosleh
ABSTRACT
Exposure to high levels of aluminum (Al) leads to neurodegeneration, which may be mediated through over-generation of free radicals. So, in the present study, we investigated the ability of both quercetin and omega 3 to ameliorate adverse effects of Al on brain antioxidants by monitoring the main brain antioxidant enzymes on molecular and cellular levels. The obtained results indicated that Al induced oxidative stress through induction of free radical production and inhibition of activity and expression of the antioxidant enzymes catalase (CAT), glutathione reductase (GR), and glutathione peroxidase (GPx); and at the same time induced superoxide dismutase (SOD) activity and gene expression. Both quercetin (QE) and omega 3 have the ability to overcome Al-induced oxidative stress, manifested by the significant reduction in free radical concentration and induction of the activity and gene expression of the brain antioxidant enzymes.
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Aluminum chloride solution, 1.0 M in nitrobenzene
Sigma-Aldrich
Aluminum chloride, reagent grade, 98%
Sigma-Aldrich
Aluminum chloride, ReagentPlus®, 99%