Merck
  • Antagonistic effects of atipamezole, yohimbine, and prazosin on xylazine-induced diuresis in clinically normal cats.
Antagonistic effects of atipamezole, yohimbine, and prazosin on xylazine-induced diuresis in clinically normal cats.
Canadian journal of veterinary research = Revue canadienne de recherche veterinaire (2014-10-31)
Yusuke Murahata, Yuya Miki, Yoshiaki Hikasa
ABSTRACT
This study aimed to investigate and compare the antagonistic effects of atipamezole, yohimbine, and prazosin on xylazine-induced diuresis in clinically normal cats. Five cats were repeatedly used in each of the 9 groups. One group was not medicated. Cats in the other groups received 2 mg/kg BW xylazine intramuscularly, and saline (as the control); 160 μg/kg BW prazosin; or 40, 160, or 480 μg/kg BW atipamezole or yohimbine intravenously 0.5 h later. Urine and blood samples were collected 10 times over 8 h. Urine volume, pH, and specific gravity; plasma arginine vasopressin (AVP) concentration; and creatinine, osmolality, and electrolyte values in both urine and plasma were measured. Both atipamezole and yohimbine antagonized xylazine-induced diuresis, but prazosin did not. The antidiuretic effect of atipamezole was more potent than that of yohimbine but not dose-dependent, in contrast to the effect of yohimbine at the tested doses. Both atipamezole and yohimbine reversed xylazine-induced decreases in both urine specific gravity and osmolality, and the increase in free water clearance. Glomerular filtration rate, osmolar clearance, and plasma electrolyte concentrations were not significantly altered. Antidiuresis of either atipamezole or yohimbine was not related to the area under the curve for AVP concentration, although the highest dose of both atipamezole and yohimbine increased plasma AVP concentration initially and temporarily, suggesting that this may in part influence antidiuretic effects of both agents. The diuretic effect of xylazine in cats may be mediated by α2-adrenoceptors but not α1-adrenoceptors. Atipamezole and yohimbine can be used as antagonistic agents against xylazine-induced diuresis in clinically normal cats.
Product Description
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Lidocaine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Xylazine hydrochloride, ≥99.0% (HPLC)
Sigma-Aldrich
Yohimbine hydrochloride, ≥98% (HPLC), powder
USP
Lidocaine, United States Pharmacopeia (USP) Reference Standard
Xylazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Yohimbine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Prazosin hydrochloride, ≥99.0% (HPLC)
USP
Prazosin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Atipamezole, ≥98% (HPLC)
Prazosin hydrochloride, European Pharmacopoeia (EP) Reference Standard