Merck
  • Insights into mitochondrial fatty acid synthesis from the structure of heterotetrameric 3-ketoacyl-ACP reductase/3R-hydroxyacyl-CoA dehydrogenase.
Insights into mitochondrial fatty acid synthesis from the structure of heterotetrameric 3-ketoacyl-ACP reductase/3R-hydroxyacyl-CoA dehydrogenase.
Nature communications (2014-09-10)
Rajaram Venkatesan, Shiv K Sah-Teli, Luqman O Awoniyi, Guangyu Jiang, Piotr Prus, Alexander J Kastaniotis, J Kalervo Hiltunen, Rik K Wierenga, Zhijun Chen
ABSTRACT
Mitochondrial fatty acid synthesis (mtFAS) is essential for respiratory growth in yeast and mammalian embryonic survival. The human 3-ketoacyl-acyl carrier protein (ACP) reductase (KAR) of mtFAS is a heterotetrameric α2β2-assembly composed of 17β-hydroxysteroid dehydrogenase type-8 (HSD17B8, α-subunit) and carbonyl reductase type-4 (CBR4, β-subunit). Here we provide a structural explanation for the stability of the heterotetramer from the crystal structure with NAD(+) and NADP(+) bound to the HSD17B8 and CBR4 subunits, respectively, and show that the catalytic activity of the NADPH- and ACP-dependent CBR4 subunit is crucial for a functional HsKAR. Therefore, mtFAS is NADPH- and ACP dependent, employing the 3R-hydroxyacyl-ACP intermediate. HSD17B8 assists in the formation of the competent HsKAR assembly. The intrinsic NAD(+)- and CoA-dependent activity of the HSD17B8 subunit on the 3R-hydroxyacyl-CoA intermediates may indicate a role for this subunit in routing 3R-hydroxyacyl-CoA esters, potentially arising from the metabolism of unsaturated fatty acids, into the mitochondrial β-oxidation pathway.
Product Description
Supelco
1,4-Benzoquinone, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
p-Benzoquinone, for spectrophotometric det. of amines, ≥99.5% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
p-Benzoquinone, reagent grade, ≥98%
Pricing and availability is not currently available.
USP
1,4-Benzoquinone, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.