Merck
  • Comparison of hollow fiber liquid-phase microextraction and ultrasound-assisted low-density solvent dispersive liquid-liquid microextraction for the determination of drugs of abuse in biological samples by gas chromatography-mass spectrometry.
Comparison of hollow fiber liquid-phase microextraction and ultrasound-assisted low-density solvent dispersive liquid-liquid microextraction for the determination of drugs of abuse in biological samples by gas chromatography-mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-03-25)
Liang Meng, Wenwen Zhang, Pinjia Meng, Binling Zhu, Kefang Zheng
ABSTRACT
Two microextraction techniques based on hollow fiber liquid-phase microextraction (HF-LPME) and ultrasound-assisted low-density solvent dispersive liquid-liquid microextraction (UA-LDS-DLLME) had been applied for the determination of drugs of abuse (methamphetamine, amphetamine, 3,4-methylenedioxymethamphetamine, 3,4-methylenedioxyamphetamine, methcathinone, ketamine, meperidine, and methadone) in urine and blood samples by gas chromatography-mass spectrometry. Parameters affecting extraction efficiency have been investigated and optimized for both methods. Under the optimum conditions, linearities were observed for all analytes in the range 0.0030-10 μg/ml with the correlation coefficient (R) ranging from 0.9985 to 0.9995 for HF-LPME and in the range 0.0030-10 μg/ml with the R ranging from 0.9985 to 0.9994 for DLLME. The recovery of 79.3-98.6% with RSDs of 1.2-4.5% was obtained for HF-LPME, and the recovery of 79.3-103.4% with RSDs of 2.4-5.7% was obtained for DLLME. The LODs (S/N=3) were estimated to be in the range from 0.5 to 5 ng/ml and 0.5 to 4 ng/ml, respectively. Compared with HF-LPME, the UA-LDS-DLLME technique had the advantages of less extraction time, suitability for batches of sample pretreatment simultaneously, and higher extraction efficiency, while HF-LPME has excellent sample clean-up effect, and is a robust and suitable technique for various sample matrices with better repeatability. Both methods were successfully applied to the analysis of drugs of abuse in real human blood sample.
Sigma-Aldrich
Rhein, technical grade
Supelco
Butyl acetate, analytical standard
Sigma-Aldrich
Butyl acetate, natural, ≥98%, FG
Sigma-Aldrich
Butyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Butyl acetate, anhydrous, ≥99%
Sigma-Aldrich
Rhein
Supelco
Butyl acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Butyl acetate, suitable for HPLC, 99.7%
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Butyl acetate, ReagentPlus®, 99.5%
Sigma-Aldrich
Butyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Cyclohexanone, ACS reagent, ≥99.0%
Sigma-Aldrich
Cyclohexanone, ReagentPlus®, 99.8%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Cyclohexanone, puriss. p.a., ≥99.5% (GC)
Supelco
Cyclohexanone, Selectophore, ≥99.5%
Sigma-Aldrich
Cyclohexanone, 99.8%
Sigma-Aldrich
Octyl acetate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Octyl acetate, ≥98%, FCC, FG
Sigma-Aldrich
Octyl acetate, ≥99%
Supelco
Cyclohexanone, analytical standard
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%