Merck
Molecular engineering of manipulated alginate-based polyurethanes.
Carbohydrate polymers (2014-08-19)
Hamed Daemi, Mehdi Barikani
ABSTRACT
The novel soluble alginate-based polyurethanes in organic solvents were synthesized by the reaction of NCO-terminated prepolymers and tributylammonium alginate (TBA-Alg) for the first time. The chemical structures of synthesized polyurethanes were characterized using FTIR, (1)H NMR and TGA. The reaction completion was confirmed by disappearing of NCO band in FTIR spectra. Furthermore, a peak at 4.71 ppm and some small peaks at a range of 4.12-4.37 ppm in the (1)H NMR of alginate-based polyurethanes were assigned to the backbone of alginate. The results of both FTIR and (1)H NMR were remarkably confirmed by TGA data. The ionic nature of polyurethane backbone not only affects on thermal properties of samples, but it also changes the chemically-bonded alginate morphology. Both polyether and polyester based non-ionic polyurethanes extended by TBA-Alg illustrated the distinct alginate, whereas those ionomers extended by alginate were appeared as the continuous systems at nanoscale.
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, suitable for HPLC, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ACS reagent, ≥99.0%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, biotech. grade, ≥99.7%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Supelco
N,N-Dimethylformamide, analytical standard
Supelco
1-Methyl-2-pyrrolidinone, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Tributylamine, puriss. plus, ≥99.5% (GC)
Sigma-Aldrich
Isophorone diisocyanate, 98%, mixture of isomers
Sigma-Aldrich
Tributylamine, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
Dimethyl sulfoxide, analytical standard