Merck
  • Rapid synthesis of 4-arylchromenes from ortho-substituted alkynols: A versatile access to restricted isocombretastatin A-4 analogues as antitumor agents.
Rapid synthesis of 4-arylchromenes from ortho-substituted alkynols: A versatile access to restricted isocombretastatin A-4 analogues as antitumor agents.
European journal of medicinal chemistry (2014-12-22)
Dolor Renko, Olivier Provot, Evelia Rasolofonjatovo, Jérôme Bignon, Jordi Rodrigo, Joëlle Dubois, Jean-Daniel Brion, Abdallah Hamze, Mouad Alami
ABSTRACT
Potent anticancer 4-arylchromene agents 6, as restricted isoCA-4 analogues, were prepared with excellent yields by a rapid and versatile synthetic pathway. First, in the presence of PTSA in EtOH, a variety of arylalkynols 9 were transformed into substituted 4-chromanones 10 in a one pot procedure which include regioselective arylalkynols hydration, alcohol etherification, MOM-cleavage, and cyclization. Further palladium coupling reactions, using aryl halides and N-tosylhydrazones 11 gave access to a small library of functionalized 4-arylchromenes 6 with good yields. From this series of 4-arylchromenes, we have identified compound 6s which inhibit tubulin assembly at a micromolar level and demonstrate a remarkable nanomolar level of cytotoxicity against four human cancer cell lines. Docking studies showed that isoCA-4 and its restricted chromene analogue 6s adopt a similar positioning in the colchicine binding-site of tubulin.
Product Description
Supelco
Vanillin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Vanillin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Vanillin, European Pharmacopoeia (EP) Reference Standard
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
p-Anisaldehyde, 98%
Sigma-Aldrich
p-Anisaldehyde, ≥97.5%, FCC, FG
Supelco
p-Anisaldehyde, analytical standard
Sigma-Aldrich
p-Anisaldehyde, natural, FG