Merck
  • Exposure assessment of 3-monochloropropane-1, 2-diol esters from edible oils and fats in China.
Exposure assessment of 3-monochloropropane-1, 2-diol esters from edible oils and fats in China.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-12-03)
Chang Li, Shao-Ping Nie, Yong-Qiang Zhou, Ming-Yong Xie
ABSTRACT
3-monochoropropane-1, 2-diol (3-MCPD) esters from edible oils are considered to be a possible risk factor for adverse effects in human. In the present study, the exposure assessment of 3-MCPD esters to Chinese population was performed. A total of 143 edible oil and fat samples collected from Chinese markets were determined for the concentrations of 3-MCPD esters. The concentration data together with the data of fats consumed were analyzed by the point evaluation and probabilistic assessment for the exposure assessment. The point evaluation showed that the mean daily intake (DI) of 3-MCPD esters were lower than the value of provisional maximum tolerable daily intake (PMTDI) of 3-MCPD (2 µg/kg BW/d). The mean DI values in different age groups obtained from probabilistic assessment were similar to the results of the point evaluation. However, in high percentiles (95th, 97.5th, 99th), the DI values in all age groups were undesirably higher than the value of PMTDI. Overall, the children and adolescents exposed more to 3-MCPD esters than the adults. Uncertainty was also analyzed for the exposure assessment. Decreasing the level of 3-MCPD esters in edible oils and consuming less oil were top priority to minimize the risk of 3-MCPD esters.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
(±)-3-Chloro-1,2-propanediol, 98%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Supelco
(±)-3-Chloro-1,2-propanediol, analytical standard
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%