Merck
  • Nonmarrow hematopoiesis occurs in a hyaluronic-acid-rich node and duct system in mice.
Nonmarrow hematopoiesis occurs in a hyaluronic-acid-rich node and duct system in mice.
Stem cells and development (2014-06-11)
Sunhee Hwang, Seung J Lee, Sang H Park, Brahmananda R Chitteti, Edward F Srour, Scott Cooper, Giao Hangoc, Hal E Broxmeyer, Byoung S Kwon
ABSTRACT
A hyaluronic-acid-rich node and duct system (HAR-NDS) was found on the surface of internal organs of mice, and inside their blood and lymph vessels. The nodes (HAR-Ns) were filled with immune cells of the innate system and were especially enriched with mast cells and histiocytes. They also contained hematopoietic progenitor cells (HPCs), such as granulocyte-macrophage, erythroid, multipotential progenitors, and mast cell progenitors (MCPs). MCPs were the most abundant among the HPCs in HAR-Ns. Their frequency was fivefold higher than that of the MCPs in bone marrow. In addition, the system contained pluripotent stem cells (PSCs) capable of producing CD45(-)Flk1(+) hemangioblast-like cells, which subsequently generated various types of HPCs and differentiated blood cells. Although HAR-Ns did not appear to harbor enough number of cells capable of long-term reconstitution or short-term radioprotection of lethally irradiated recipients, bone marrow cells were able to engraft in the HAR-NDS and reconstitute hematopoietic potentials of the system. PSCs and HPCs were consistently found in intravenous, intralymphatic, and intestinal HAR-ND. We infer that PSCs and HPCs reside in the HAR-ND and that this novel system may serve as an alternative means to traffic immature and mature blood cells throughout the body.
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Interleukin-7 from mouse, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
EPO human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
SCF from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Stem Cell Factor from mouse, SCF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Erythropoietin human, EPO, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine