Merck
  • Ambiguanine A-G, hexahydrobenzophenanthridine alkaloids from Corydalis ambigua var. amurensis.
Ambiguanine A-G, hexahydrobenzophenanthridine alkaloids from Corydalis ambigua var. amurensis.
Phytochemistry (2014-06-12)
Zhi-You Yang, Bao-Hong Jiang, Fu-Kang Teng, Na Han, Zhi-Hui Liu, Jing-Jing Zhou, Yu Wang, De-An Guo, Jun Yin
ABSTRACT
Seven hexahydrobenzophenanthridine-type alkaloids, Ambiguanine A-G, along with eight known alkaloids, were isolated from tubers of Corydalis ambigua var. amurensis. Their structures were elucidated based on extensive spectroscopic analyses, with absolute configurations determined by CD experiments.
Product Description
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>