Merck
  • Knockdown of GALNT1 suppresses malignant phenotype of hepatocellular carcinoma by suppressing EGFR signaling.
Knockdown of GALNT1 suppresses malignant phenotype of hepatocellular carcinoma by suppressing EGFR signaling.
Oncotarget (2015-03-03)
Miao-Juei Huang, Rey-Heng Hu, Chih-Hsing Chou, Chia-Lang Hsu, Ya-Wen Liu, John Huang, Ji-Shiang Hung, I-Rue Lai, Hsueh-Fen Juan, Sung-Liang Yu, Yao-Ming Wu, Min-Chuan Huang
ABSTRACT
O-glycosylation is a common protein modification. Aberrant O-glycosylation is associated with many cancers. GALNT1 is a GalNAc-transferase that initiates protein O-glycosylation. We found that GALNT1 is frequently up-regulated in hepatocellular carcinoma (HCC) and is associated with poor patient survival. Overexpression of GALNT1 increased and knockdown decreased HCC cell migration and invasion. Knockdown of GALNT1 inhibited EGF-induced migration and invasion. Knockdown of GALNT1 decreased EGFR activation and increased EGFR degradation, by decreasing EGFR O-glycosylation. This study demonstrates that down-regulation of GALNT1 is sufficient to suppress malignant phenotype of HCC cells by decreasing EGFR signaling. Thus, GALNT1 is a potential target in HCC.
Sigma-Aldrich
EGF from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
EGF from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human GALNT1
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Galnt1
Sigma-Aldrich
EGF from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)