Merck
  • Genome-wide shRNA screen revealed integrated mitogenic signaling between dopamine receptor D2 (DRD2) and epidermal growth factor receptor (EGFR) in glioblastoma.
Genome-wide shRNA screen revealed integrated mitogenic signaling between dopamine receptor D2 (DRD2) and epidermal growth factor receptor (EGFR) in glioblastoma.
Oncotarget (2014-03-25)
Jie Li, Shan Zhu, David Kozono, Kimberly Ng, Diahnn Futalan, Ying Shen, Johnny C Akers, Tyler Steed, Deepa Kushwaha, Michael Schlabach, Bob S Carter, Chang-Hyuk Kwon, Frank Furnari, Webster Cavenee, Stephen Elledge, Clark C Chen
ABSTRACT
Glioblastoma remains one of the deadliest of human cancers, with most patients succumbing to the disease within two years of diagnosis. The available data suggest that simultaneous inactivation of critical nodes within the glioblastoma molecular circuitry will be required for meaningful clinical efficacy. We conducted parallel genome-wide shRNA screens to identify such nodes and uncovered a number of G-Protein Coupled Receptor (GPCR) neurotransmitter pathways, including the Dopamine Receptor D2 (DRD2) signaling pathway. Supporting the importance of DRD2 in glioblastoma, DRD2 mRNA and protein expression were elevated in clinical glioblastoma specimens relative to matched non-neoplastic cerebrum. Treatment with independent si-/shRNAs against DRD2 or with DRD2 antagonists suppressed the growth of patient-derived glioblastoma lines both in vitro and in vivo. Importantly, glioblastoma lines derived from independent genetically engineered mouse models (GEMMs) were more sensitive to haloperidol, an FDA approved DRD2 antagonist, than the premalignant astrocyte lines by approximately an order of magnitude. The pro-proliferative effect of DRD2 was, in part, mediated through a GNAI2/Rap1/Ras/ERK signaling axis. Combined inhibition of DRD2 and Epidermal Growth Factor Receptor (EGFR) led to synergistic tumoricidal activity as well as ERK suppression in independent in vivo and in vitro glioblastoma models. Our results suggest combined EGFR and DRD2 inhibition as a promising strategy for glioblastoma treatment.
Product Description
Haloperidol for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Risperidone, ≥98% (HPLC), powder
Haloperidol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-741,626, ≥98% (HPLC)
Sigma-Aldrich
Spiperone, solid
Sigma-Aldrich
Haloperidol, powder
Risperidone, European Pharmacopoeia (EP) Reference Standard
USP
Risperidone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
MISSION® esiRNA, targeting human GNAI2
USP
Haloperidol, United States Pharmacopeia (USP) Reference Standard
Haloperidol for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Gnai2
Sigma-Aldrich
MISSION® esiRNA, targeting human DRD2
Risperidone for system suitability, European Pharmacopoeia (EP) Reference Standard