Merck
  • Exposure to di-2-ethylhexyl phthalate, di-n-butyl phthalate and bisphenol A through infant formulas.
Exposure to di-2-ethylhexyl phthalate, di-n-butyl phthalate and bisphenol A through infant formulas.
Journal of agricultural and food chemistry (2015-03-03)
Teresa Cirillo, Giuseppe Latini, Maria Antonietta Castaldi, Lucia Dipaola, Evelina Fasano, Francesco Esposito, Gelsomina Scognamiglio, Fabio Di Francesco, Luigi Cobellis
ABSTRACT
Phthalates and bisphenol A (BPA) are ubiquitous contaminants identified as endocrine disruptors. Phthalates are worldwide used as plasticizers, in particular to improve the mechanical properties of polymers such as polyvinyl chloride. Because they are not chemically bound to the polymer, they tend to leach out with time and use. Di-2-ethylhexyl phthalate (DEHP) and di-n-butyl phthalate (DnBP) are the two most common phthalates. BPA is an estrogenic compound used to manufacture polycarbonate containers for food and drink, including baby bottles. It can migrate from container into foods, especially at elevated temperatures. Diet is a predominant source of exposure for phthalates and BPA, especially for infants. The aim of this study was to test the presence of DEHP, DnBP, and BPA in infant formulas. DEHP, DnBP, and BPA concentrations were measured in 22 liquid and 28 powder milks by gas chromatography with flame ionization detection and high performance liquid chromatography with fluorimetric detection, respectively. DEHP concentrations in our samples were between 0.005 and 5.088 μg/g (median 0.906 μg/g), DnBP concentrations were between 0.008 and 1.297 μg/g (median 0.053 μg/g), and BPA concentrations were between 0.003 and 0.375 μg/g (median 0.015 μg/g). Concentrations of the investigated contaminants in liquid and powder milks were not significantly different, even though samples were packed in different types of containers. These data point out potential hazards for infants fed with baby formulas. Contamination seems more related to the production of formulas than to a release from containers.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%