Merck
  • Alloy Cu₃Pt nanoframes through the structure evolution in Cu-Pt nanoparticles with a core-shell construction.
Alloy Cu₃Pt nanoframes through the structure evolution in Cu-Pt nanoparticles with a core-shell construction.
Scientific reports (2014-09-19)
Lin Han, Hui Liu, Penglei Cui, Zhijian Peng, Suojiang Zhang, Jun Yang
ABSTRACT
Noble metal nanoparticles with hollow interiors and customizable shell compositions have immense potential for catalysis. Herein, we present an unique structure transformation phenomenon for the fabrication of alloy Cu₃Pt nanoframes with polyhedral morphology. This strategy starts with the preparation of polyhedral Cu-Pt nanoparticles with a core-shell construction upon the anisotropic growth of Pt on multiply twinned Cu seed particles, which are subsequently transformed into alloy Cu₃Pt nanoframes due to the Kirkendall effect between the Cu core and Pt shell. The as-prepared alloy Cu₃Pt nanoframes possess the rhombic dodecahedral morphology of their core-shell parents after the structural evolution. In particular, the resulting alloy Cu₃Pt nanoframes are more effective for oxygen reduction reaction but ineffective for methanol oxidation reaction in comparison with their original Cu-Pt core-shell precursors.
Product Description
SAFC
Oleylamine, ≥98% (primary amine)
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Chloroplatinic acid hexahydrate, powder and chunks
Sigma-Aldrich
Chloroplatinic acid hexahydrate, ACS reagent, ≥37.50% Pt basis
Sigma-Aldrich
Chloroplatinic acid hexahydrate, BioXtra