Merck
  • Immunoregulation of dendritic cells by the receptor T cell Ig and mucin protein-3 via Bruton's tyrosine kinase and c-Src.
Immunoregulation of dendritic cells by the receptor T cell Ig and mucin protein-3 via Bruton's tyrosine kinase and c-Src.
Journal of immunology (Baltimore, Md. : 1950) (2014-08-31)
Neeraj Maurya, Ravindra Gujar, Mamta Gupta, Vinod Yadav, Saurabh Verma, Pradip Sen
ABSTRACT
The receptor T cell Ig and mucin protein-3 (TIM-3) has emerged as an important regulator of innate immune responses. However, whether TIM-3-induced signaling promotes or inhibits the activation and maturation of dendritic cells (DCs) still remains uncertain. In addition, the TIM-3 signaling events involved in this immunoregulatory function are yet to be established. In this article, we report that TIM-3 crosslinking by anti-TIM-3 Ab inhibited DC activation and maturation by blocking the NF-κB pathway. After Ab-mediated crosslinking, TIM-3 became tyrosine phosphorylated, which then sequentially bound and activated the nonreceptor tyrosine kinases Bruton's tyrosine kinase (Btk) and c-Src. Activation of Btk-c-Src signaling in turn triggered the secretion of some inhibitory factor (or factors) from DCs that inhibited the NF-κB pathway and subsequent activation and maturation of DCs. Silencing of Btk or c-Src abrogated the inhibitory effects of TIM-3 on DCs. These results demonstrate an essential role for Btk-c-Src signaling in TIM-3-induced DC suppression. Thus, in addition to demonstrating an inhibitory role for TIM-3 signaling in DC activation, we define the molecular mechanism by which TIM-3 mediates this effect.
Sigma-Aldrich
MISSION® esiRNA, targeting human BTK
Sigma-Aldrich
DL-Tyrosine, 99%
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Btk