Merck
Synthesis and antibacterial properties of pyrimidine derivatives.
Acta poloniae pharmaceutica (2015-04-09)
Jerzy Cieplik, Marcin Stolarczyk, Janusz Pluta, Olaf Gubrynowicz, Iwona Bryndal, Tadeusz Lis, Marcin Mikulewicz
ABSTRACT
In this study, a series of syntheses was conducted on the pyrimidine system, obtaining bisulfite carboxyl derivatives 4 and hydroxy derivatives 5. In addition, a series of syntheses were carried out as a result of which both alkyl and aromatic amines were obtained. Then, the attempt was made to cyclize these amines in the Mannich reaction to pyrimido[4,5-d]pyrimidines 11, 12. After determination of chemical structure using physicochemical tests, also by means of crystallographic tests, all the newly obtained derivatives underwent microbiological tests on bacterial strains and fungi. The most interesting results of the microbiological tests are included later in the study.
Product Description
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Erythromycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard