Merck
  • Acute toxicity and genotoxicity study of fermented traditional herb formula Guibi-tang.
Acute toxicity and genotoxicity study of fermented traditional herb formula Guibi-tang.
Journal of ethnopharmacology (2014-09-15)
Hwayong Park, Youn-Hwan Hwang, Hye Jin Yang, Hyun-Kyu Kim, Kyung Seuk Song, Jin Yeul Ma
ABSTRACT
Guibi-tang (Guipi-tang in Chinese and Kihi-to in Japanese) is a multi-herb traditional medicine commonly prescribed to treat psychoneurosis in East Asia. Although this medicine has been widely used, there is little available information on the safety and toxicity of Guibi-tang, especially on the fermented one. Guibi-tang, composed of 12 herbs, was fermented with bacteria and lyophilized. Single dose acute toxicity in rats was observed for 14 days after administration. Genetic toxicity of fermented Guibi-tang was evaluated on bacterial reverse mutation in Salmonella and Escherichia spp., chromosome aberrations in Chinese hamster ovary cells, and micronucleus formation in mice. Ingredients in FGBT were identified and quantified by high performance liquid chromatography-mass spectrometry. In acute oral toxicity study, behavior, clinical signs and body weight changes were normal observing in all experimental animals. No revertant colonies were found in any bacterial cultures examined. Morphological or numerical anomalies and significant increased number of aberrant metaphases were not observed. Micronucleus assay showed no significant increases in the frequency of inducing micronuclei in any dose examined. Decursinol, decursin, glycyrrhizin, and 6-gingerol in fermented Guibi-tang were identified and quantitated. As a whole, no acute and genotoxic effects were found in all the assays and parameters analyzed. Fermented Guibi-tang was recognized as safe and non-toxic, and therefore can be used for applications of traditional medicine in modern complementary and alternative therapeutics and health care.
Sigma-Aldrich
9-Aminoacridine hydrochloride monohydrate, 98%
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Acetic acid, analytical standard
Supelco
Tetrachloroethylene, analytical standard
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Supelco
[6]-Gingerol, analytical standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice), ≥95.0% (NT)
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Supelco
Glycyrrhizic acid ammonium salt, analytical standard, suitable for HPLC
Sigma-Aldrich
Tetrachloroethylene, ACS reagent, ≥99.0%
Ammonium glycyrrhizate, European Pharmacopoeia (EP) Reference Standard
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland