Merck
  • Measuring the chemical and cytotoxic variability of commercially available kava (Piper methysticum G. Forster).
Measuring the chemical and cytotoxic variability of commercially available kava (Piper methysticum G. Forster).
PloS one (2014-11-05)
Amanda C Martin, Ed Johnston, Chengguo Xing, Adrian D Hegeman
ABSTRACT
Formerly used world-wide as a popular botanical medicine to reduce anxiety, reports of hepatotoxicity linked to consuming kava extracts in the late 1990s have resulted in global restrictions on kava use and have hindered kava-related research. Despite its presence on the United States Food and Drug Administration consumer advisory list for the past decade, export data from kava producing countries implies that US kava imports, which are not publicly reported, are both increasing and of a fairly high volume. We have measured the variability in extract chemical composition and cytotoxicity towards human lung adenocarcinoma A549 cancer cells of 25 commercially available kava products. Results reveal a high level of variation in chemical content and cytotoxicity of currently available kava products. As public interest and use of kava products continues to increase in the United States, efforts to characterize products and expedite research of this potentially useful botanical medicine are necessary.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Ethyl acetate
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%