Merck
  • Efficient human immunodeficiency virus (HIV-1) infection of cells lacking PDZD8.
Efficient human immunodeficiency virus (HIV-1) infection of cells lacking PDZD8.
Virology (2015-03-17)
Shijian Zhang, Joseph Sodroski
ABSTRACT
PDZD8 can bind the capsid proteins of different retroviruses, and transient knockdown of PDZD8 results in a decrease in the efficiency of an early, post-entry event in the retrovirus life cycle. Here we used the CRISPR-CAS9 system to create cell lines in which PDZD8 expression is stably eliminated. The PDZD8-knockout cell lines were infected by human immunodeficiency virus (HIV-1) and murine leukemia virus as efficiently as the parental PDZD8-expressing cells. These results indicate that PDZD8 is not absolutely necessary for HIV-1 infection and diminishes its attractiveness as a potential target for intervention.
Sigma-Aldrich
MISSION® esiRNA, targeting human PDZD8
Sigma-Aldrich
Anti-PDZD8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pdzd8