Merck
  • Increased Toll-like receptor 5 expression indicates esophageal columnar dysplasia.
Increased Toll-like receptor 5 expression indicates esophageal columnar dysplasia.
Virchows Archiv : an international journal of pathology (2013-11-14)
Olli Helminen, Heikki Huhta, Heikki Takala, Petri P Lehenkari, Juha Saarnio, Joonas H Kauppila, Tuomo J Karttunen
ABSTRACT
Toll-like receptor 5 (TLR5) is an immune receptor, which recognizes bacterial flagellin. Increased expression has been reported in various premalignant and malignant lesions indicating a role in carcinogenesis. We assessed the expression of TLR5 in normal esophageal squamous epithelium, Barrett's esophagus with and without dysplasia, and in esophageal adenocarcinoma. Specimens with normal esophagus (n=93), gastric (n=75) or intestinal metaplasia (n=53) without dysplasia, and low-grade (n=56) or high-grade dysplasia (n=33) and esophageal adenocarcinoma (n=94) were studied. TLR5 immunohistochemical stainings were analyzed for the proportion of positive cells and the intensity of expression. In normal squamous epithelium, only the basal third showed TLR5 expression. In esophageal gastric or intestinal metaplasia, expression was present in majority of the cells but significantly weaker (p<0.001) than in dysplastic epithelium. In dysplasia, expression extended to the apical cytoplasm, contrasting basolateral expression in non-dysplastic columnar epithelium. Receiver operating characteristic curve analysis showed that moderate to high expression intensity of TLR5 indicates low-grade dysplasia with 86 % sensitivity and 83 % specificity. Carcinomas showed increased expression in comparison with non-dysplastic columnar epithelium, but there was no association with prognosis. Our results indicate that the esophageal columnar dysplasia is associated with clear increase of TLR5 expression and dissolution of regular polarized expression. TLR5 staining provides a possible biomarker for the recognition of low-grade dysplasia. In addition, the findings suggest a role for abnormal expression of TLR5 in the pathogenesis of esophageal adenocarcinoma and suggest importance of altered microbiome in the pathogenesis of complications of Barrett's esophagus.