- Expression of leukocyte-associated immunoglobulin-like receptor-1 (LAIR-1) on osteoclasts and its potential role in rheumatoid arthritis.
Expression of leukocyte-associated immunoglobulin-like receptor-1 (LAIR-1) on osteoclasts and its potential role in rheumatoid arthritis.
Leukocyte-associated immunoglobulin-like receptor-1 is an inhibitory receptor primarily expressed by immune cells. This study was undertaken to define the role of this molecule in osteoclast differentiation and rheumatoid arthritis. In vitro osteoclast assays were performed to characterize the role of Leukocyte-associated immunoglobulin-like receptor-1 in murine and human osteoclastogenesis. Human Leukocyte-associated immunoglobulin-like receptor-1 expression was assessed by immunohistochemistry staining in the synovium of patients with rheumatoid arthritis. The levels of soluble Human Leukocyte-associated immunoglobulin-like receptor-1 were determined by enzyme-linked immunosorbent assay. We found that multinucleated osteoclast formation from mouse bone marrow cells was inhibited by treatment with a monoclonal antibody against mouse Leukocyte-associated immunoglobulin-like receptor-1 in vitro. By immunohistochemistry, we found that Leukocyte-associated immunoglobulin-like receptor-1 was mainly expressed by macrophages in the inflamed synovial tissue of rheumatoid arthritis patients. In addition, serum and synovial fluid levels of soluble Leukocyte-associated immunoglobulin-like receptor-1 were higher in rheumatoid arthritis patients compared to healthy controls or osteoarthritis patients. Moreover, overexpression of Leukocyte-associated immunoglobulin-like receptor-1 in CD14+ monocytes from healthy volunteers also inhibited human osteoclastogenesis. Collectively, these data demonstrate for the first time that Leukocyte-associated immunoglobulin-like receptor-1 inhibits osteoclastogenesis. Therefore, these results may have therapeutic implications for the treatment of rheumatoid arthritis.