Merck
  • A Korean patient with glutaric aciduria type 1 with a novel mutation in the glutaryl CoA dehydrogenase gene.
A Korean patient with glutaric aciduria type 1 with a novel mutation in the glutaryl CoA dehydrogenase gene.
Annals of clinical and laboratory science (2014-05-06)
Hee Su Kim, Hee Joon Yu, Jeehun Lee, Hyung-Doo Park, Ji Hye Kim, Hyung-Jin Shin, Dong Kyu Jin, Munhyang Lee
ABSTRACT
Mutations in the glutaryl-CoA dehydrogenase gene can result in Glutaric aciduria type 1(GA 1) by accumulation of glutaric acid, 3-hydroxyglutaric acid (3-OH-GA), and glutarylcarnitine (C5DC). GA 1 is characterized by macrocephaly, subdural hemorrhage (SDH), and dystonic movement disorder after acute encephalopathic crisis. We report a Korean patient with GA1 and a novel mutation. A 16-month-old boy presented with SDH, macrocephaly, and developmental delay. In the neurologic examination, the patient had mild axial hypotonia, but otherwise normal neurologic functions. The brain MRI showed large amounts of bilateral SDH and high signal intensity in both basal ganglia and thalamus. Metabolic screening tests detected highly elevated urinary GA levels but 3-OH-glutaric acid was normal. C5DC was 0.94 μM/L (reference range < 0.3 μM/L). The patient had compound heterozygous mutations of the GCDH gene: p.Arg257Gln (c.770G>A) and p.Cys308Arg (c.922T>C). p.Cys308Arg is a novel mutation; reports of p.Arg257Gln were also rare both in Caucasians and Asian populations. In summary, we hereby report one Korean patient with GA1 with clinical, biochemical, and radiologic characteristics confirmed by genetic analysis.
Sigma-Aldrich
Glutaryl-L-carnitine lithium salt, ≥98.0% (TLC)