Merck
  • Cobalt electrolyte/dye interactions in dye-sensitized solar cells: a combined computational and experimental study.
Cobalt electrolyte/dye interactions in dye-sensitized solar cells: a combined computational and experimental study.
Journal of the American Chemical Society (2012-11-02)
Edoardo Mosconi, Jun-Ho Yum, Florian Kessler, Carlos J Gómez García, Cristiano Zuccaccia, Antonio Cinti, Mohammad K Nazeeruddin, Michael Grätzel, Filippo De Angelis
ABSTRACT
We report a combined experimental and computational investigation to understand the nature of the interactions between cobalt redox mediators and TiO(2) surfaces sensitized by ruthenium and organic dyes, and their impact on the performance of the corresponding dye-sensitized solar cells (DSSCs). We focus on different ruthenium dyes and fully organic dyes, to understand the dramatic loss of efficiency observed for the prototype Ru(II) N719 dye in conjunction with cobalt electrolytes. Both N719- and Z907-based DSSCs showed an increased lifetime in iodine-based electrolyte compared to the cobalt-based redox shuttle, while the organic D21L6 and D25L6 dyes, endowed with long alkoxy chains, show no significant change in the electron lifetime regardless of employed electrolyte and deliver a high photovoltaic efficiency of 6.5% with a cobalt electrolyte. Ab initio molecular dynamics simulations show the formation of a complex between the cobalt electrolyte and the surface-adsorbed ruthenium dye, which brings the [Co(bpy)(3)](3+) species into contact with the TiO(2) surface. This translates into a high probability of intercepting TiO(2)-injected electrons by the oxidized [Co(bpy)(3)](3+) species, lying close to the N719-sensitized TiO(2) surface. Investigation of the dye regeneration mechanism by the cobalt electrolyte in the Marcus theory framework led to substantially different reorganization energies for the high-spin (HS) and low-spin (LS) reaction pathways. Our calculated reorganization energies for the LS pathways are in excellent agreement with recent data for a series of cobalt complexes, lending support to the proposed regeneration pathway. Finally, we systematically investigate a series of Co(II)/Co(III) complexes to gauge the impact of ligand substitution and of metal coordination (tris-bidentate vs bis-tridentate) on the HS/LS energy difference and reorganization energies. Our results allow us to trace structure/property relations required for further development of cobalt electrolytes for DSSCs.
Sigma-Aldrich
FK 209 Co(III) PF6 salt
Sigma-Aldrich
FK 102 Co(III) TFSI salt
Sigma-Aldrich
FK 102 Co(II) PF6 salt
Sigma-Aldrich
FK 102 Co(III) PF6 salt
Sigma-Aldrich
FK 209 Co(II) TFSI salt
Sigma-Aldrich
FK 209 Co(III) TFSI salt