Merck
  • Functional cloning and characterization of a novel nonhomeodomain protein that inhibits the binding of PBX1-HOX complexes to DNA.
Functional cloning and characterization of a novel nonhomeodomain protein that inhibits the binding of PBX1-HOX complexes to DNA.
The Journal of biological chemistry (2000-05-29)
C Abramovich, W F Shen, N Pineault, S Imren, B Montpetit, C Largman, R K Humphries
ABSTRACT
PBX1 is a homeodomain protein that functions in complexes with other homeodomain-containing proteins to regulate gene expression during developmental and/or differentiation processes. A yeast two-hybrid screen of a fetal liver-hematopoietic cDNA library using PBX1a as bait led to the discovery of a novel non-homeodomain-containing protein that interacts with PBX1 as well as PBX2 and PBX3. RNA analysis revealed it to be expressed in CD34(+) hematopoietic cell populations enriched in primitive progenitors, as is PBX1; search of the expressed sequence tag data base indicated that it is also expressed in other early embryonic as well as adult tissues. The full-length cDNA encodes a 731-amino acid protein that has no significant homology to known proteins. This protein that we have termed hematopoietic PBX-interacting protein (HPIP) is mainly localized in the cytosol and in small amounts in the nucleus. The region of PBX that interacts with HPIP includes both the homeodomain and immediate N-terminal flanking sequences. Strikingly, electrophoretic mobility shift assays revealed that HPIP inhibits the ability of PBX-HOX heterodimers to bind to target sequences. Moreover, HPIP strongly inhibits the transactivation activity of E2A-PBX. Together these findings suggest that HPIP is a new regulator of PBX function.